Learn More
Waters Corp Waters Corporation Ultrastyragel Column, 10000 Å, 5 μm, 19 mm × 300 mm, Toluene
Supplier: Waters Corp WAT025869
This Ultrastyragel column provides high-resolution preparative GPC for efficient isolation and sample cleanup. It consolidates tasks typically requiring multiple smaller columns into a single, more efficient unit. Featuring a 10000 Å pore size, 5 μm particle size, and measuring 19 mm inner diameter by 300 mm length, it is packed in Toluene and supports a molecular weight range of 5000 to 600000.
- GPC chemistry with SEC/GPC separation mode.
- Polymer particle substrate, spherical shape.
- pH range of 2 to 12.
- Molecular weight range of 5000 to 600000.
- Particle size of 5 μm and pore size of 10000 Å.
- Column format, packed in Toluene.
- Compatible with HPLC systems for SEC technique.
- Inner diameter of 19 mm and length of 300 mm.
- One unit per package.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.