Learn More
Waters Corp Waters GPC Column, 500 Angstrom, 5 micrometer, 7.8 mm, 300 mm
Supplier:  Waters Corp WAT085501
Encompass_Preferred
Waters GPC Column, 500 Angstrom, 5 micrometer, 7.8 mm, 300 mm
Catalog No. 50-785-940