Learn More
Waters Corp Waters Ultrastyragel Column, 5 μm, 19 mm, 500 Å
Supplier: Waters Corp WAT025860
This product is designed for high resolution preparative GPC applications.
- Particle size: 5 μm
- Inner diameter: 19 mm
- Length: 300 mm
- Particle substrate: Polymer
- Pore size: 500 Å
- Molecular weight range: 100 to 10000
- pH range: 2 to 12 pH
- Separation mode: SEC/GPC
- Technique: SEC
- System: HPLC
- Packing solvent: THF
- Particle shape: Spherical
- Contains hazardous material requiring special freight handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.