Learn More
Selleck Chemical LLC UM-164 5mg 903564-48-7
SDP
Supplier:  Selleck Chemical LLC S87065MG
Encompass_Preferred
UM-164 is a highly potent, dual c-Src/p38inhibitor of c-Src with a binding constant Kd of 2.7 nM for c-Src and inhibits both p38? and p38?. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8308