Learn More
Medchemexpress LLC HY-N0573 100mg Medchemexpress, Umbelliferone CAS:93-35-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0573100MG
Encompass_Preferred
Medchemexpress, HY-N0573 100mg Umbelliferone CAS:93-35-6 Umbelliferone, a natural product of the coumarin family, is a fluorescing compound which can be used as a sunscreen agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0288