Learn More
Medchemexpress LLC Umbelliferone (7-Hydroxycoumarin) | 93-35-6 | MFCD00006764 | 162.14 | 1 G
SDP
Supplier:  Medchemexpress LLC HYN05731G
Encompass_Preferred
Umbelliferone (7-Hydroxycoumarin) is a natural, orally active coumarin family product that functions as a fluorescing compound and can be used as a sunscreen agent. It induces cell cycle arrest, apoptosis, and DNA fragmentation in HepG2 cells. It also exhibits significant anticancer effects, attenuates allergic airway inflammation, displays neuroprotective effects, crosses the blood-brain barrier, and shows anti-inflammatory and antioxidant effects in chronic alcohol-fed rats.
  • Induces cell cycle arrest, apoptosis, and DNA fragmentation in HepG2 cells.
  • Exhibits significant anticancer effects.
  • Attenuates allergic airway inflammation.
  • Displays neuroprotective effects and crosses the blood-brain barrier.
  • Shows anti-inflammatory and antioxidant properties.
Catalog No. 50-003-88494