Learn More
Medchemexpress LLC Umbelliferone-d5 | 1215373-23-1 | 167.17 | 2.5 MG
SDP
Supplier:  Medchemexpress LLC HYN0573S2.5MG
Encompass_Preferred
Umbelliferone-d5 (7-Hydroxycoumarin-d5) is a deuterium-labeled coumarin derivative, a natural and orally active compound. It is a fluorescing agent and has shown diverse biological activities including anticancer, anti-inflammatory, and neuroprotective effects. This compound serves as a valuable tool for quantitative analysis in research.
  • Deuterium-labeled for quantitative analysis
  • Can be used as a tracer in drug development
  • Acts as an internal standard for techniques like NMR, GC-MS, or LC-MS
  • Exhibits anticancer effects by inducing cell cycle arrest and apoptosis
  • Attenuates allergic airway inflammation
  • Displays neuroprotective, anti-inflammatory, and antioxidant properties
Catalog No. 50-003-88496