Learn More
Medchemexpress LLC Umbelliprenin | 23838-17-7 | 99.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY127029A1MG
Encompass_Preferred
Umbelliprenin is a prenylated coumarin that acts as an antioxidant with anticancer, anti-inflammatory, and immunomodulatory effects. It inhibits lipoxygenase, which helps manage inflammation pathways. In inflammatory macrophages, Umbelliprenin suppresses nitric oxide production and inducible nitric oxide synthase (NO synthase).
  • Acts as an antioxidant
  • Possesses anticancer effects
  • Has anti-inflammatory effects
  • Exhibits immunomodulatory properties
  • Inhibits lipoxygenase to manage inflammation pathways
  • Suppresses nitric oxide production in inflammatory macrophages
  • Alleviates neuropathic pain in in vivo studies
  • Decreases serum IL-6 levels and oxidative stress
Catalog No. 50-003-40866