Learn More
Medchemexpress LLC Umbelliprenin | 23838-17-7 | 99.9% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY127029A10MG
Encompass_Preferred
Umbelliprenin is a prenylated coumarin that acts as an antioxidant. It exhibits anticancer, anti-inflammatory, and immunomodulatory effects. It functions by inhibiting lipoxygenase, thereby managing inflammation pathways. In inflammatory macrophages, it suppresses nitric oxide production and inducible nitric oxide synthase (NO synthase).
  • Acts as an antioxidant
  • Exhibits anticancer effects
  • Exhibits anti-inflammatory effects
  • Exhibits immunomodulatory effects
  • Functions by inhibiting lipoxygenase
  • Suppresses nitric oxide production
  • Suppresses inducible nitric oxide synthase (NO synthase)
Catalog No. 50-003-42986