Learn More
Medchemexpress LLC Umbelliprenin | 23838-17-7 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY127029A
Encompass_Preferred
Umbelliprenin is a prenylated coumarin and an antioxidant. It exhibits anticancer, anti-inflammatory, and immunomodulatory properties. This compound functions as a lipoxygenase inhibitor, contributing to the management of inflammatory pathways. Studies indicate that Umbelliprenin can suppress nitric oxide production and inducible nitric oxide synthase (NO synthase) in inflammatory macrophages, highlighting its potential in modulating immune responses.
  • Antioxidant properties
  • Exhibits anticancer effects
  • Anti-inflammatory properties
  • Immunomodulatory effects
  • Inhibits lipoxygenase
  • Suppresses nitric oxide production
  • Modulates inducible nitric oxide synthase in inflammatory macrophages
  • Suitable for research use
Catalog No. 50-003-40098