Learn More
Medchemexpress LLC Umbelliprenin | 23838-17-7 | 99.9% | 366.49 g·mol-1 | C24H30O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY127029A5MG
Encompass_Preferred
Umbelliprenin is a prenylated coumarin research compound with antioxidant, anticancer, anti-inflammatory, and immunomodulatory activities. It inhibits lipoxygenase and suppresses inducible nitric oxide synthase in inflammatory macrophages, making it useful for biochemical and cellular studies of inflammation and related pathways.
  • Inhibits lipoxygenase and suppresses inducible nitric oxide synthase.
  • Useful for studies of inflammation, immunomodulation, and metabolic enzyme pathways.
  • High reported purity suitable for analytical and biological assays.
  • Supplied in small vial quantities convenient for screening and mechanistic assays.
  • Stable when stored protected from light at -20°C; in solvent, store at -80°C for up to 6 months, or at -20°C for up to 1 month.
Catalog No. 50-002-63971