Learn More
Medchemexpress LLC Umbralisib R-enantiomer | 1532533-69-9 | 99.5% | 50 MG

Supplier: Medchemexpress LLC HY12279F50MG
Umbralisib R-enantiomer is the R-stereoisomer of the PI3Kδ inhibitor Umbralisib, supplied for research use. It is the less active enantiomer and shows selectivity for the δ isoform over α, β, and γ. The compound is supplied as a solid, white to light yellow powder, and is soluble in DMSO.
- Research use only.
- Less active R-enantiomer of Umbralisib.
- Selective PI3Kδ inhibitor with reduced activity versus α, β, and γ isoforms.
- Purity 99.5%.
- Molecular weight 571.55 g/mol; formula C31H24F3N5O3.
- Soluble in DMSO (33.33 mg/mL); ultrasonic assistance recommended.
- Packaged as 50 MG.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.