Learn More
Medchemexpress LLC HY-12100 5mg Medchemexpress, Umeclidinium (bromide) CAS:869113-09-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY121005MG
Encompass_Preferred
Medchemexpress, HY-12100 5mg Umeclidinium (bromide) CAS:869113-09-7 Umeclidinium bromide is a novel mAChR antagonist. The affinity (Ki) of Umeclidinium bromide for the cloned human M1-M5 mAChRs ranges from 0.05 to 0.16 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0959