Learn More
Medchemexpress LLC Umeclidinium bromide | 869113-09-7 | 99.1% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1210050MG
Encompass_Preferred
Umeclidinium bromide is a potent muscarinic acetylcholine receptor antagonist used for receptor pharmacology and binding studies. It is supplied as a white to off-white solid appropriate for analytical and research applications.
  • Potent mAChR antagonist with Ki 0.05-0.16 nM.
  • High purity suitable for analytical and assay applications.
  • Chemical formula C29H34BrNO2; molecular weight 508.49.
  • Available in small lab pack sizes for research use.
Catalog No. 50-002-33994