Learn More
Medchemexpress LLC Umibecestat | 1387560-01-1 | 99.8% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11968950MG
Encompass_Preferred
Umibecestat is a small-molecule beta-secretase (BACE-1) inhibitor used for research into Alzheimer's disease. Provided as a research-grade solid for in vitro and preclinical studies, it has formula C19H15ClF7N5O2 and a molecular weight of 513.8 g/mol.
  • Beta-secretase (BACE-1) inhibitor for Alzheimer's disease research.
  • High purity suitable for biochemical and cellular assays.
  • Available in multiple pack sizes, including 50 MG solid vials.
  • Also available as a 10 mM solution in DMSO for ready-to-use experiments.
Catalog No. 50-002-33327