Learn More
Unchained Labs UNA, 10 KDA MWCO, CASE OF 200
SDP
Supplier:  Unchained Labs 9012011
Encompass
Una consumable for Unagi instrument, 0.5 - 8 mL capacity.MWCO: 10kDa, regenerated cellulose (RC). 8 Unas per box,25 boxes, case total of 200
Catalog No. NC2210889
Only null left
Add to Cart
Request a Quote