Learn More
STA PHARMACEUTICAL US LLC UNA 2'-OBz G(iBu) amidite | 5 g | CAS 1120329-61-4 | InChIKey PCZJDFIZVYBABW-WESQYFQMSA-N
Supplier: STA PHARMACEUTICAL US LLC STAAM10156AK1005GM
UNA 2'-OBz G(iBu) amidite is a Amidite reagent (Subcategory: UNA Series) sold by WuXi TIDES. Offered in 5 g. Store at -20 °C. SDS available for reference.
Specifications
- CAS: 1120329-61-4
- MDL: No data
- InChIKey: PCZJDFIZVYBABW-WESQYFQMSA-N
- Molecular Weight: 962.053761998
- Molecular Formula: C51H60N7O10P
- Purity: ≥95%
- Container Type: 100 mL Glass (Septum)
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 100.4 g
- Commodity Code: 29335995
- Country Of Origin: China
- IUPAC: (2R)-2-(((2R)-1-(bis(4-methoxyphenyl)(phenyl)methoxy)-3-(((2-cyanoethoxy)(diisopropylamino)phosphaneyl)oxy)propan-2-yl)oxy)-2-(2-isobutyramido-6-oxo-1,6-dihydro-9H-purin-9-yl)ethyl benzoate
- SMILES: CC(N(P(OC[C@H](O[C@@H](N1C(N=C(N2)NC(C(C)C)=O)=C(N=C1)C2=O)COC(C3=CC=CC=C3)=O)COC(C4=CC=C(C=C4)OC)(C5=CC=C(C=C5)OC)C6=CC=CC=C6)OCCC#N)C(C)C)C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.