Learn More
Perkin Elmer US LLC Un-Assembled Aluminum Crimp Cap with Aluminum/Silicone Septa and Star Spring, 1000/pk
Supplier:  Perkin Elmer US LLC B0104244
Encompass_Preferred
For sealing sample vials for temperatures up to 220°C. Very low background peaks, very low permeability, not usable for repetitive injections from the same vial.
Catalog No. 50-211-2353