Learn More
Medchemexpress LLC Unc 0631 | 1320288-19-4 | 99.3% | C37H61N7O2 | 10MG

Supplier: Medchemexpress LLC HY1380810MG
UNC 0631 is a potent, research-grade inhibitor of the histone methyltransferase G9a. It inhibits G9a enzymatic activity (biochemical IC50 = 4 nM) and reduces H3K9me2 levels in multiple cell lines at low nanomolar concentrations, making it useful for epigenetics and chromatin-modification research.
- Potent G9a inhibition (biochemical IC50 = 4 nM).
- Effective cellular reduction of H3K9me2 at low nanomolar concentrations (examples: MCF7 18 nM; MDA-MB-231 25 nM).
- High purity for analytical applications (99.3%).
- Stable solid with recommended storage: powder -20°C for long term, 4°C for short term.
- Supplied as a light yellow crystalline solid suitable for research use.
- Suitable for studies of chromatin modification and epigenetic regulation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.