Learn More
Medchemexpress LLC UNC-2170 50mg | 1648707-58-7 | 50 MG

Supplier: Medchemexpress LLC HY11553150MG
UNC-2170 is a fragment-like small-molecule ligand for 53BP1, a methyl-lysine reader protein involved in DNA damage response. It binds 53BP1 with micromolar affinity and is used as a biochemical probe for studying 53BP1 interactions and function.
- Functionally active fragment-like ligand for 53BP1.
- Binds 53BP1 with Kd = 22 μM and inhibits with IC50 = 29 μM.
- Shows at least 17-fold selectivity for 53BP1 versus other methyl-lysine reader proteins.
- Molecular weight 313.23; formula C14H21BrN2O.
- Available as solid powder or as DMSO solution for biochemical assays.
- Recommended storage: solid at 4°C; solutions at -80°C (up to 6 months) or -20°C (up to 1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.