Learn More
Selleck Chemical LLC UNC 3230-E0305-5MG
SDP
Supplier:  Selleck Chemical LLC E03055MG
Encompass_Preferred
UNC3230 is a potent selective and ATP-competitive PIP5K1C inhibitor with an IC50 of 41 nM and also inhibits PIP4K2C It also has antinociceptive and anticancer effects
Catalog No. 50-313-8291