Learn More
Medchemexpress LLC UNC-CA2-103 | 99.3% | 344.37 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY16242650MG
Encompass_Preferred
UNC-CA2-103 is a TLK2 inhibitor with a human IC50 of 18 nM. It demonstrates high selectivity over TLK1 and most other kinases, making it suitable for research in various cancer types.
  • Potently inhibits purified TLK2 enzyme with an IC50 of 18 nM.
  • Exhibits narrow kinome selectivity.
  • Applicable in research concerning breast cancer, glioblastoma, and gastric cancer.
Catalog No. 50-003-92100