Learn More
Medchemexpress LLC UNC-CA2-103 | 99.3% | 344.37 g·mol⁻¹ | C20H16N4O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY16242610MG
Encompass_Preferred
UNC-CA2-103 is a small-molecule research compound that potently inhibits Tousled-like kinase 2 (TLK2) with a reported IC50 of 18 nM. It is supplied as a powder and as a DMSO solution, has a molecular weight of 344.37 g·mol⁻¹, and is provided for laboratory research use only.
  • Potent TLK2 inhibition (IC50 18 nM) for biochemical and cellular studies.
  • High purity suitable for analytical and experimental applications.
  • Available in multiple package sizes and a ready-to-use DMSO solution for assay flexibility.
  • Stable when stored under recommended conditions to maintain compound integrity.
  • Comes with product documentation describing handling and storage recommendations.
Catalog No. 50-002-88496