Learn More
Medchemexpress LLC HY-12335 5mg Medchemexpress, UNC0379 CAS:1620401-82-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY123355MG
Encompass_Preferred
Medchemexpress, HY-12335 5mg UNC0379 CAS:1620401-82-2 UNC0379 is a selective, substrate-competitive inhibitor of lysine methyltransferase SETD8 (KMT5A) with an IC50 of 7.3 μM; selective over 15 other methyltransferases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9957
 Restricted Item