Learn More
Selleck Chemical LLC UNC0638 10mg 1255580-76-7
SDP
Supplier:  Selleck Chemical LLC S807110MG
Encompass_Preferred
UNC0638 is a potent, selective and cell-penetrant chemical probe for G9a and GLP histone methyltransferase with IC50 of <15 nM and 19 nM, respectively, shows selectivity over a wide range of epigenetic and non-epigenetic targets. UNC0638 has anti-viral activities. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5627