Learn More
Selleck Chemical LLC UNC0642 5mg 1481677-78-4
SDP
Supplier:  Selleck Chemical LLC S72305MG
Encompass_Preferred
UNC0642 is a potent, selective inhibitor of histone methyltransferases G9a/GLP with IC50s less than 2.5 nM for G9a and GLP and shows more than 300-fold selective for G9a and GLP over a broad range of kinases, GPCRs, transporters, and ion channels. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8098