Learn More
Cayman Chemical UNC1999, 1431612-23-5, 5 mg
SDP
Supplier:  Cayman Chemical 146215
Encompass_Preferred
A selective, cell-permeable EZH2 inhibitor (IC50 = 2 nM) that has been shown to inhibit H3K27methylation in MCF10A cells with an IC50 value of 124 nM
Catalog No. NC0616595