Learn More
Selleck Chemical LLC UNC2025 S9662-100MG
SDP
Supplier:  Selleck Chemical LLC S9662100MG
Encompass_Preferred
UNC2025 is a potent and orally active dual inhibitor of FLT3 and MER with IC50 of 0 35 nM and 0 46 nM respectively UNC2025 also inhibits AXL TRKA TRKC QIK TYRO3 SLK NuaK1 Kit (c-Kit) and Met (c-Met) with IC50 of 1 65 nM 1 67 nM 4 38 nM 5 75 nM 5 83 nM 6 14 nM 7 97 nM 8 18 nM and 364 nM respectively
Catalog No. 50-000-24610