Learn More
Apexbio Technology LLC UNC2025 1429881-91-3 5mg
SDP
Supplier:  Apexbio Technology LLC B80165
Encompass_Preferred
UNC2025 (CAS 1429881-91-3) is a potent orally bioavailable dual inhibitor targeting the receptor tyrosine kinases Mer and Flt3 It demonstrates low nanomolar inhibitory activity against Mer (IC50 0 8 nM) and Flt3 (IC50 0 74 nM) with selective activity over related kinases such as Axl and Tyro3 In biochemical and cellular assays UNC2025 effectively inhibits Mer phosphorylation in B-ALL and Flt3 phosphorylation in Flt3-ITD positive AML models resulting in reduced colony formation in relevant cell lines In xenograft mouse models oral administration substantially decreased Mer phosphorylation in leukemic cells UNC2025 is valuable for probing Mer and Flt3 signaling in preclinical oncological research
Catalog No. 50-000-88043