Learn More
Selleck Chemical LLC UNC2025 HCl
SDP
Supplier:  Selleck Chemical LLC S75765MG
Encompass_Preferred
UNC2025 HCl is a potent and orally bioavailable dual MER/FLT3 inhibitor with IC50 of 0 74 nM and 0 8 nM respectively about 20-fold selectivity over Axl and Tyro3
Catalog No. 50-000-04750