Learn More
Medchemexpress LLC UNC2025 hydrochloride | 2070015-17-5 | MFCD28963885 | 100.0% | 513.12 g/mol | C28H41ClN6O | 5MG
SDP
Supplier:  Medchemexpress LLC HY12344A5MG
Encompass_Preferred
UNC2025 hydrochloride is a small-molecule, ATP-competitive inhibitor of Mer and FLT3 kinases with low-nanomolar potency and favorable pharmacokinetic properties; it is provided for research use in studies of Mer/FLT3 signaling and leukemia models.
  • Potent Mer/FLT3 inhibitor with IC50 values in the sub-nanomolar to low-nanomolar range.
  • Demonstrates selectivity for MERTK relative to Axl.
  • High reported purity suitable for research applications.
  • Solid, light yellow to yellow physical form for easy handling and storage.
  • Manufacturer-reported solubility in DMSO, water, and PBS for formulation flexibility.
  • Storage recommendations provided for stability in solution and solid state.
Catalog No. 50-202-9959