Learn More
Selleck Chemical LLC UNC2250 5mg 1493694-70-4
SDP
Supplier:  Selleck Chemical LLC S73425MG
Encompass_Preferred
UNC2250 is a potent and selective Mer inhibitor with IC50 of 1.7 nM, about 160- and 60-fold selectivity over the closely related kinases Axl/Tyro3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1075106