Learn More
Apexbio Technology LLC UNC2881 1493764-08-1 50mg
SDP
Supplier:  Apexbio Technology LLC B435750
Encompass_Preferred
UNC2881 (CAS 1493764-08-1) is a selective inhibitor of the Mer receptor tyrosine kinase a member of the TAM (Tyro3 Axl Mer) subfamily involved in processes such as platelet aggregation and macrophage activation In biochemical assays UNC2881 demonstrates over 50% inhibition of MER and several kinases at 430 nM while in 697 B-ALL cells it inhibits Mer phosphorylation with an IC50 of 22 nM UNC2881 also suppresses ligand-dependent phosphorylation of Mer-EGFR chimeric proteins and inhibits collagen-induced platelet aggregation and ATP release in human plasma With oral bioavailability of 14% in mice and high systemic clearance UNC2881 serves as a valuable tool for investigating Mer-mediated signaling in thrombosis and related pathophysiological contexts
Catalog No. 50-000-88506