Learn More
Medchemexpress LLC Unc569 | 1350547-65-7 | 99.9% | 396.50 g/mol | C22H29FN6 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11759625MG
Encompass_Preferred
Unc569 is a potent, reversible, ATP-competitive Mer receptor tyrosine kinase inhibitor used for preclinical oncology research. It exhibits low-nanomolar potency against Mer and activity against Axl and Tyro3, and is supplied as a high-purity solid or DMSO solution for biochemical and cellular studies.
  • High purity suitable for research use.
  • Potent Mer inhibition (IC50 2.9 nM).
  • Also inhibits Axl and Tyro3, enabling TAM-family studies.
  • Available as solid and DMSO solution formats for assay flexibility.
  • Intended for in vitro and in vivo preclinical research.
Catalog No. 50-002-27802