Learn More
Medchemexpress LLC UNC6852 | 2688842-08-0 | 99.6% | 832.97 g/mol | C43H48N10O6S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1307085MG
Encompass_Preferred
UNC6852 is a PROTAC degrader that targets the EED subunit of the polycomb repressive complex 2 (PRC2). It is provided as a high-purity research compound with documented solubility and storage recommendations for both in vitro and in vivo applications.
  • Targets the EED subunit of PRC2 for selective protein degradation.
  • High purity suitable for biochemical and cellular studies.
  • Supplied as a solid with light yellow to yellow appearance.
  • Soluble in DMSO at high concentration; in vivo formulations documented.
  • Stable with recommended storage conditions for powder and solution.
Catalog No. 50-002-63011