Learn More
Medchemexpress LLC UNC7467 | 2922283-43-8 | C20H13NO3 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY150607
Encompass_Preferred
UNC7467 is a potent IP6K inhibitor designed for use in obesity research. It effectively reduces levels of inositol pyrophosphates and has demonstrated the ability to ameliorate diet-induced obesity, insulin resistance, and hepatic steatosis in mouse models.
  • Potent IP6K inhibitor
  • Reduces levels of inositol pyrophosphates
  • Useful for obesity research
  • Ameliorates diet-induced obesity
  • Improves insulin resistance
  • Reduces hepatic steatosis in diet-induced obesity mice
Catalog No. 50-003-74386