Learn More
Aviva Systems Biology Corp UNC84A Antibody - N-terminal region (ARP49929_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49929_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UNC84A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human UNC84A. Alternate names: FLJ12407, KIAA0810, MGC176649, SUN1, UNC84AApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60218