Learn More
Medchemexpress LLC Unc9995 | 1354030-52-6 | 99.9% | 10 MG

Supplier: Medchemexpress LLC HY12092010MG
UNC9995 is a β-arrestin2-biased agonist of the dopamine D2 receptor (Drd2), supplied as a white to off-white solid for laboratory research use only. It inhibits NLRP3 inflammasome activation by enhancing β-arrestin2-NLRP3 interaction and has been used in preclinical studies to modulate inflammation and depressive-like behaviors.
- β-arrestin2-biased agonist of dopamine D2 receptor
- CAS 1354030-52-6; molecular formula C20H21Cl2N3OS
- Purity 99.9%
- Molecular weight 422.37 g·mol⁻1
- Soluble in DMSO (50 mg/mL with warming and sonication)
- Store powder at -20°C; store solutions at -80°C for long term
- For research use only; not for human or clinical use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.