Learn More
Cayman Chemical Uncaa yncopyla sutn 10mg
SDP
Supplier:  Cayman Chemical 3493910 MG
Encompass_Preferred
An indole alkaloid; reduces glutamate-induced cytotoxicity in primary rat cerebellar granule neurons from 100-300 µM; inhibits nicotine-induced dopamine release in PC12 cells from 1-100 µM
Catalog No. 50-291-0544