Learn More
ATCC Uncinocarpus reesii Sigler et Orr, freeze-dried fungal type strain (UAMH 3880) - ATCC 34533
Supplier:  ATCC 34533
Encompass
Uncinocarpus reesii Sigler et Orr (ATCC 34533) is a freeze-dried filamentous fungal type strain (UAMH 3880) provided as a research and reference culture. It is classified as Biosafety Level 1 and was originally isolated from feathers in Australia. The product is supplied lyophilized with detailed rehydration and inoculation instructions to aid reliable recovery.
  • Freeze-dried format for extended shelf life and convenient storage.
  • Type strain with verified strain designation UAMH 3880, suitable for taxonomic and comparative studies.
  • BSL-1 classification appropriate for standard laboratory containment.
  • Recommended growth on PYG medium at 24°C to support robust recovery and growth.
  • Includes stepwise rehydration and inoculation instructions for reliable revival of culture.
Catalog No. NC3308299