Learn More
Vector Laboratories Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), Unconjugated | 2mg
SDP
Supplier:  Vector Laboratories L11702
Encompass
Tomato lectin (from Lycopersicon esculentum) is an effective marker of blood vessels and microglial cells in rodents. Conjugation of the lectin with a fluorophore facilitates fast, one-step detection and visualization using intravascular perfusion methods or direct application to tissue sections.
Catalog No. NC1770389