Learn More
ACROBiosystems Unconjugated Human HLA-A0201
Supplier:  ACROBiosystems HLMH52H6200UG
Encompass_Preferred
Unconjugated Human HLA-A*0201B2MMAGE-A4 (GVYDGREHTV) Complex Protein (HLM-H52H6) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-A*0201) Ile 21 - Met 119 (B2M) GVYDGREHTV peptide (Accession # AAA59606.1 (HLA-A*0201) P61769-1 (B2M) GVYDGREHTV).
Catalog No. 50-253-4721