Learn More
Medchemexpress LLC Undecane | 1120-21-4 | 100 G
Supplier: Medchemexpress LLC HYN8593100G
Undecane is a potent cAMP agonist with anti-allergic and anti-inflammatory properties. It inhibits degranulation and the secretion of histamine and TNF-α. Undecane also reverses elevated levels of p38 phosphorylation, NF-κB transcriptional activity, and the expression of target cytokine/chemokine genes such as thymus and activation-regulated chemokine (TARC), macrophage-derived chemokine (MDC), and interleukin-8 (IL-8). This compound is suitable for research into skin inflammatory disorders, including atopic dermatitis.
- Potent cAMP agonist
- Anti-allergic and anti-inflammatory activities
- Inhibits degranulation and the secretion of histamine and TNF-α
- Reverses increased levels of p38 phosphorylation and NF-κB transcriptional activity
- Modulates target cytokine/chemokine genes (TARC, MDC, IL-8)
- Useful for studying skin inflammatory disorders, including atopic dermatitis