Learn More
Medchemexpress LLC Undecanedioic acid | 1852-04-6 | MFCD00004444 | 98.0% | 216.28 g/mol | C11H20O4 | 50 G

Supplier: Medchemexpress LLC HYW01412550G
Undecanedioic acid is an endogenous dicarboxylic acid (C11H20O4, MW 216.28 g/mol) supplied as a solid research standard with confirmed purity. It is intended for analytical, biochemical, and metabolic studies, and includes formulation and storage guidance for solution and in vivo preparations.
- Purity ≥98.0% for analytical use.
- White to off-white solid appearance.
- Soluble in DMSO (100 mg/mL); ultrasonic assistance may be required.
- Recommended storage: 4°C sealed; in solvent: -80°C (6 months), -20°C (1 month).
- Available in multiple pack sizes including 50 g.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.