Learn More
Med Vet International Reusable Underpad Beck's Classic 34 X 36 Inch Polyester / Rayon Moderate Absorbency, 7136HB, Each
Supplier:  Med Vet International 1123368EA
Encompass_Preferred
Beck's reusable underpads have a soft cotton/polyester top layer The knitted barrier and vinyl backing prevent liquids from staining furniture, bedding and mattresses A quilted soaker made of a polyester/rayon blend draws moisture away from the body Beck's classic underpads are washable for repeat use They come in size 34- x 36-inch and a range of colors to suit your preferences
Catalog No. 50-304-4794