Learn More
ARROW POINT PARTNERS INC UNI-SEP MAXI 50ML 10ML 18PK
Supplier:  ARROW POINT PARTNERS INC U10
Encompass_Preferred
Blood Separation Tube, Pre-filled w/10 ml separation medium, UNI-SEP, Maxi, 50 ml plas tube, Pre-filled w/10 ml separation medium, 5.6% polysucrose 9.6% sodium metrizoate, Ready-to-use, One-way barrier, 4.50 HI, 1.00 DIA, w/Rack, 18 tubes per Bx
Catalog No. 50-157-345
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.