Learn More
Medchemexpress LLC HY-B0873 500mg Medchemexpress, Uniconazole CAS:83657-22-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0873500MG
Encompass_Preferred
Medchemexpress, HY-B0873 500mg Uniconazole CAS:83657-22-1 Uniconazole is a plant growth regulator that functions by inhibiting cytochrome P450 707As (Ki=68 nM), a family of enzymes that catabolize Abscisic acid, and thus, suppress gibberellin and sterol biosynthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2669