Learn More
Cayman Chemical UnIdentIfIed AcremIn I 1mg
SDP
Supplier:  Cayman Chemical 297711 MG
Encompass_Preferred
A fungal metabolite; inhibits germination of P. viticola sporangia by 23.2 and 32.8% at 0.5 and 1 mM, respectively
Catalog No. 50-299-0227