Learn More
Cayman Chemical UnIdentIfIed Quadron 2.5mg
SDP
Supplier:  Cayman Chemical 266882.5 MG
Encompass_Preferred
A sesquiterpene originally isolated from A. terreus; cytotoxic to human carcinoma KB cells in vitro (EC50 = 1.3 μg/ml)
Catalog No. 50-293-4178