Learn More
Microsolv Technology Corporation Union, Connector Kit, High Pressure, 0.50mm Through Hole with 10-32 Screw Threads. Stainless Steel, Straight Line. Use with 1/16" Tubing. 1 EA.
Supplier:  Microsolv Technology Corporation 4940503
Encompass_Preferred
This low dead volume, "Union" is used to connect the same size PEEK or Stainless Steel tubing on both ends, in the high pressures zones of HPLC systems. Supplied with Two Stainless Steel Wrench Tighten Connectors and matching Ferrules.
Catalog No. 50-227-1577